accessibility menu, dialog, popup

UserName

CAS RN 22776-09-6

IUPAC Name: 1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)ethan-1-one
Synonyms: 1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)ethanone
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: VGRSZNPWUZDPCH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2OCCCOC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results