accessibility menu, dialog, popup

UserName

CAS RN 22679-54-5

IUPAC Name: (4-tert-butylphenyl)(phenyl)methanone
Synonyms: (4-tert-butylphenyl)(phenyl)methanone
Molecular Weight (g/mol): 238.33
Molecular Formula: C17H18O
InChi Key: DFYJCXSOGSYMAJ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results