accessibility menu, dialog, popup

UserName

ICH3OONCH3CH3H3C

CAS RN 2260-50-6

IUPAC Name: [2-(acetyloxy)ethyl]trimethylazanium iodide
Synonyms: acetylcholine iodide
Molecular Weight (g/mol): 273.11
Molecular Formula: C7H16INO2
InChi Key: SMBBQHHYSLHDHF-UHFFFAOYSA-M
SMILES: [I-].CC(=O)OCC[N+](C)(C)C
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)