accessibility menu, dialog, popup

UserName

CAS RN 2257-69-4

IUPAC Name: 4-chloro-1,2-dihydrophthalazin-1-one
Synonyms: 4-chloro-2H-phthalazin-1-one
Molecular Weight (g/mol): 180.59
Molecular Formula: C8H5ClN2O
InChi Key: QCKGMJDOJRNSMS-UHFFFAOYSA-N
SMILES: ClC1=NNC(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)