accessibility menu, dialog, popup

UserName

CAS RN 2257-09-2

SCN

CAS RN 2257-09-2

IUPAC Name: (2-isothiocyanatoethyl)benzene
Synonyms: phenethyl isothiocyanate peitc β-phenethyl isothiocyanate phenylaethylsenfoel 1-phenethylisothiocyanate
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: IZJDOKYDEWTZSO-UHFFFAOYSA-N
SMILES: S=C=NCCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)