accessibility menu, dialog, popup

UserName

CAS RN 2247-91-8

H3COOCH2FFF

CAS RN 2247-91-8

IUPAC Name: 3,3,3-trifluoroprop-1-en-2-yl acetate
Synonyms: 3,3,3-trifluoroprop-1-en-2-yl acetate
Molecular Weight (g/mol): 154.09
Molecular Formula: C5H5F3O2
InChi Key: VOKGSDIHTCTXDS-UHFFFAOYSA-N
SMILES: CC(=O)OC(=C)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results