accessibility menu, dialog, popup

UserName

CAS RN 22457-89-2

IUPAC Name: {[(3Z)-4-{N-[(4-amino-2-methylpyrimidin-5-yl)methyl]formamido}-3-[(Z)-benzoylsulfanyl]pent-3-en-1-yl]oxy}phosphonic acid
Synonyms: benfotiamine 苯磷硫胺
Molecular Weight (g/mol): 466.45
Molecular Formula: C19H23N4O6PS
InChi Key: BTNNPSLJPBRMLZ-LGMDPLHJSA-N
SMILES: C\C(N(CC1=CN=C(C)N=C1N)C=O)=C(/CCOP(O)(O)=O)SC(=O)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)