CAS RN 22428-87-1
IUPAC Name:
1,4-dioxaspiro[4.5]decan-8-ol
Synonyms:
1,4-dioxaspiro[4.5]decan-8-ol
Molecular Weight (g/mol):
158.2
Molecular Formula:
C8H14O3
InChi Key:
HKQTYQDNCKMNHZ-UHFFFAOYSA-N
SMILES:
OC1CCC2(CC1)OCCO2
1
–
2
of
2
results
2
1,4-Dioxaspiro[4.5]decan-8-ol, 98%
CAS: 22428-87-1 Molecular Formula: C8H14O3 Molecular Weight (g/mol): 158.20 MDL Number: MFCD00067003 InChI Key: HKQTYQDNCKMNHZ-UHFFFAOYSA-N Synonym: 1,4-dioxaspiro 4.5 decan-8-ol,1,4-dioxa-spiro 4.5 decan-8-ol,4-hydroxycyclohexanone ethylene acetal,4-hydroxycyclohexanone monoethylene ketal,8-hydroxy-1,4-dioxaspiro 4.5 decane,pubchem20110,acmc-1chv5,4,4-ethylenedioxycyclohexanol PubChem CID: 567352 IUPAC Name: 1,4-dioxaspiro[4.5]decan-8-ol SMILES: OC1CCC2(CC1)OCCO2