accessibility menu, dialog, popup

UserName

CAS RN 22415-59-4

IUPAC Name: (oxolan-2-yl)methanol
Synonyms: tetrahydrofurfuryl alcohol
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: BSYVTEYKTMYBMK-UHFFFAOYNA-N
SMILES: OCC1CCCO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results