accessibility menu, dialog, popup

UserName

OFFH2N

CAS RN 22236-08-4

IUPAC Name: 3-(difluoromethoxy)aniline
Synonyms: 3-(difluoromethoxy)aniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: RSOFZRXRIPVBBM-UHFFFAOYSA-N
SMILES: NC1=CC(OC(F)F)=CC=C1
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results