accessibility menu, dialog, popup

UserName

ONOOFF

CAS RN 22236-07-3

IUPAC Name: 1-(difluoromethoxy)-3-nitrobenzene
Synonyms: 1-(difluoromethoxy)-3-nitrobenzene
Molecular Weight (g/mol): 189.12
Molecular Formula: C7H5F2NO3
InChi Key: NYVCZALWNPMMSQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(OC(F)F)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)