accessibility menu, dialog, popup

UserName

CAS RN 2221-12-7

IUPAC Name: 1-methyl-3-phenylimidazolidine-2,4-dione
Synonyms: 1-methyl-3-phenylimidazolidine-2,4-dione
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: YLKKRCYFGKGCKZ-UHFFFAOYSA-N
SMILES: CN1CC(=O)N(C1=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results