accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OClClH3CHN(R)(S)HO(S)HNCH3(R)O(R)OH(S)O(R)CH3OO(R)(S)OHHH

CAS RN 22189-32-8

IUPAC Name: dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride
Synonyms: spectinomycin
Molecular Weight (g/mol): 495.34
Molecular Formula: C14H36Cl2N2O12
InChi Key: DCHJOVNPPSBWHK-UXXUFHFZSA-N
SMILES: [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
Quantity 
  • (4)
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (1)
Type 
  • (4)
  • (2)
Grade 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (4)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)