accessibility menu, dialog, popup

UserName

CAS RN 22189-08-8

IUPAC Name: magnesium(2+) sulfate
Synonyms: magnesium sulfate magnesium sulfate anhydrous MgSO4 magnesiumsulfat magnesium(II) sulfate
Molecular Weight (g/mol): 120.36
Molecular Formula: MgO4S
InChi Key: CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES: [Mg++].[O-]S([O-])(=O)=O
Quantity 
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
Grade 
  • (3)
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (5)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)