CAS RN 22181-94-8
CAS RN 22181-94-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Benin, MedChemExpress
MedChemExpress Benin is a potent cytostatic drug that can be used for the treatment of generalized carcinoma of the breast.