missing translation for 'onlineSavingsMsg'
Learn More

Benin, MedChemExpress

Product Code. 16492338
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492338 5 mg 5mg
16453129 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492338 Supplier Medchem Express Supplier No. HYU00073

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Benin is a potent cytostatic drug that can be used for the treatment of generalized carcinoma of the breast.

Benin is a potent cytostatic drug that can be used for the treatment of generalized carcinoma of the breast.

Specifications

Chemical Name or Material Benin
CAS 22181-94-8
Color White
Purity Grade Notes Research
Molecular Formula C14H19N5O3S
Quantity 5 mg
Synonym Butocin Butocine
SMILES O=C(OCC)CNC(CCCCSC1=C2N=CNC2=NC=N1)=O
Molecular Weight (g/mol) 337.4
Formula Weight 337.4
Percent Purity 97.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.