accessibility menu, dialog, popup

UserName

CH3OBrBrOH3C

CAS RN 22094-18-4

IUPAC Name: 1,3-dibromo-2,2-dimethoxypropane
Synonyms: 1,3-dibromo-2,2-dimethoxypropane
Molecular Weight (g/mol): 261.94
Molecular Formula: C5H10Br2O2
InChi Key: CPAHOXOBYHMHDT-UHFFFAOYSA-N
SMILES: COC(CBr)(CBr)OC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results