accessibility menu, dialog, popup

UserName

HClCH3OCH3HN

CAS RN 2208-07-3

IUPAC Name: hydrogen ethyl ethanimidate chloride
Synonyms: hydrogen ethyl ethanimidate chloride
Molecular Weight (g/mol): 123.58
Molecular Formula: C4H10ClNO
InChi Key: WGMHMVLZFAJNOT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(C)=N
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)