CAS RN 2206-26-0
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Acetonitrile-d3, for NMR, 99.8 atom % D
CAS: 2206-26-0 Molecular Weight (g/mol): 44.07 MDL Number: MFCD00001881 InChI Key: WEVYAHXRMPXWCK-FIBGUPNXSA-N IUPAC Name: (²H₃)acetonitrile SMILES: [2H]C([2H])([2H])C#N
Acetonitrile-d3, for NMR, 99 atom % D
CAS: 2206-26-0 Molecular Formula: C2H3N Molecular Weight (g/mol): 44.07 MDL Number: MFCD00001881 InChI Key: WEVYAHXRMPXWCK-FIBGUPNXSA-N PubChem CID: 123151 SMILES: [2H]C([2H])([2H])C#N
Acetonitrile-d3, for NMR, 99.9 atom% D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
Acetonitrile-d{3}, 99.8% (Isotopic)
CAS: 2206-26-0 Molecular Formula: C2H3N Molecular Weight (g/mol): 44.07 MDL Number: MFCD00001881 InChI Key: WEVYAHXRMPXWCK-FIBGUPNXSA-N Synonym: acetonitrile-d3,2h3 acetonitrile,cd3cn,acetonitrile-2,2,2-d3,methyl-d3 cyanide,trideuteroacetonitrile,acetonitrile-d3, 99.8 atom % d,acetonitrile-d3, ≥99.8 atom % d,acetonitrile-d isotopic 5g PubChem CID: 123151 IUPAC Name: 2,2,2-trideuterioacetonitrile SMILES: [2H]C([2H])([2H])C#N
Acetonitrile-d{3}, 100% (Isotopic)
CAS: 2206-26-0 Molecular Formula: C2H3N Molecular Weight (g/mol): 44.07 MDL Number: MFCD00001881 InChI Key: WEVYAHXRMPXWCK-FIBGUPNXSA-N Synonym: acetonitrile-d3,2h3 acetonitrile,cd3cn,acetonitrile-2,2,2-d3,methyl-d3 cyanide,trideuteroacetonitrile,acetonitrile-d3, 99.8 atom % d,acetonitrile-d3, ≥99.8 atom % d,acetonitrile-d isotopic 5g PubChem CID: 123151 IUPAC Name: 2,2,2-trideuterioacetonitrile SMILES: [2H]C([2H])([2H])C#N
Acetonitrile-d3, for NMR, packaged in 1.00 ml ampoules, 99.95 atom % D
Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D
CAS: 2206-26-0 Molecular Formula: C2H3N Molecular Weight (g/mol): 44.07 MDL Number: MFCD00001881 InChI Key: WEVYAHXRMPXWCK-FIBGUPNXSA-N Synonym: acetonitrile-d3,2h3 acetonitrile,cd3cn,acetonitrile-2,2,2-d3,methyl-d3 cyanide,trideuteroacetonitrile,acetonitrile-d3, 99.8 atom % d,acetonitrile-d3, ≥99.8 atom % d,acetonitrile-d isotopic 5g PubChem CID: 123151 IUPAC Name: 2,2,2-trideuterioacetonitrile SMILES: [2H]C([2H])([2H])C#N
Acetonitrile-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Acetonitrile-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Acetonitrile-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.