CAS RN 22031-64-7
CAS RN 22031-64-7
IUPAC Name:
(2E)-3-phenylprop-2-enamide
Synonyms:
cinnamamide
Molecular Weight (g/mol):
147.18
Molecular Formula:
C9H9NO
InChi Key:
APEJMQOBVMLION-VOTSOKGWSA-N
SMILES:
NC(=O)\C=C\C1=CC=CC=C1
1
–
2
of
2
results
1
trans-Cinnamamide, TRC
CAS: 22031-64-7 Molecular Formula: C9 H9 N O Molecular Weight (g/mol): 147.17 Synonym: 2-Propenamide, 3-phenyl-, (2E)-,2-Propenamide, 3-phenyl-, (E)-,Cinnamamide, (E)- (8CI),(2E)-3-Phenyl-2-propenamide,(E)-3-Phenyl-2-propenamide,(E)-Cinnamamide,U 2422,trans-3-Phenyl-2-propenamide,trans-Cinnamamide,trans-Cinnamic acid amide,trans-Cinnamoylamine,trans-β-(Aminocarbonyl)styrene,Mitomycin Imp. A (EP) IUPAC Name: (E)-3-phenylprop-2-enamide SMILES: NC(=O)\C=C\c1ccccc1
2
(E)-3-Phenylprop-2-enamide (trans-Cinnamamide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.