accessibility menu, dialog, popup

UserName

CAS RN 220227-98-5

OFFFOH

CAS RN 220227-98-5

IUPAC Name: 4-hydroxy-3-(trifluoromethyl)benzaldehyde
Synonyms: 4-hydroxy-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: ZLMUFCNWTPSOSN-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C=O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results