accessibility menu, dialog, popup

UserName

CAS RN 220141-70-8

NHOHNOOOHFH2O

CAS RN 220141-70-8

IUPAC Name: 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate
Synonyms: 5-fluoroorotic acid hydrate
Molecular Weight (g/mol): 192.1
Molecular Formula: C5H5FN2O5
InChi Key: LODRRYMGPWQCTR-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=C(F)C(=O)NC(=O)N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)