accessibility menu, dialog, popup

UserName

CAS RN 22013-33-8

IUPAC Name: 2,3-dihydro-1,4-benzodioxin-6-amine
Synonyms: 2,3-dihydro-1,4-benzodioxin-6-amine
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: BZKOZYWGZKRTIB-UHFFFAOYSA-N
SMILES: NC1=CC=C2OCCOC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)