accessibility menu, dialog, popup

UserName

H3COFFF

CAS RN 219998-30-8

IUPAC Name: 1,3,5-trifluoro-2-methoxybenzene
Synonyms: 1,3,5-trifluoro-2-methoxybenzene
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: ZLRIBYUOFAKHEF-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results