accessibility menu, dialog, popup

UserName

H3COBOHHOCl

CAS RN 219735-99-6

IUPAC Name: (2-chloro-4-methoxyphenyl)boronic acid
Synonyms: 2-chloro-4-methoxyphenylboronic acid
Molecular Weight (g/mol): 186.4
Molecular Formula: C7H8BClO3
InChi Key: LOCGPWGCRVKCFN-UHFFFAOYSA-N
SMILES: COC1=CC=C(B(O)O)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results