accessibility menu, dialog, popup

UserName

CAS RN 219555-98-3

CH3NNCl

CAS RN 219555-98-3

IUPAC Name: 2-chloro-5-propylpyrimidine
Synonyms: 2-chloro-5-propylpyrimidine
Molecular Weight (g/mol): 156.61
Molecular Formula: C7H9ClN2
InChi Key: WVPHBBSAPVBUGZ-UHFFFAOYSA-N
SMILES: CCCC1=CN=C(Cl)N=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)