CAS RN 21886-62-4
CAS RN 21886-62-4
IUPAC Name:
1-(4-tert-butylphenyl)-2-chloroethan-1-one
Synonyms:
1-(4-tert-butylphenyl)-2-chloroethanone
Molecular Weight (g/mol):
210.7
Molecular Formula:
C12H15ClO
InChi Key:
LTHPBRNHHJIQME-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(C=C1)C(=O)CCl
1
–
1
of
1
results
1
4'-tert-Butyl-2-chloroacetophenone, 97%, Thermo Scientific™
CAS: 21886-62-4 Molecular Formula: C12H15ClO Molecular Weight (g/mol): 210.70 MDL Number: MFCD00171621 InChI Key: LTHPBRNHHJIQME-UHFFFAOYSA-N Synonym: 4-tert-butylphenacyl chloride,2-chloro-4'-tert-butylacetophenone,1-4-tert-butylphenyl-2-chloroethanone,1-4-tert-butyl phenyl-2-chloroethanone,4-tert-butylphenacylchloride,ethanone, 2-chloro-1-4-1,1-dimethylethyl phenyl,1-4-tert-butyl-phenyl-2-chloro-ethanone,4'-tert-butyl-2-chloroacetophenone,1-4-tert-butylphenyl-2-chloroethan-1-one,2-chloro-4-tert-butylacetophenone PubChem CID: 229661 IUPAC Name: 1-(4-tert-butylphenyl)-2-chloroethanone SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)CCl