CAS RN 21864-21-1
CAS RN 21864-21-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Lithium niobium isopropoxide, 99+% (metals basis), 5% w/v in isopropanol
CAS: 21864-21-1 Molecular Formula: C18H42LiNbO6 MDL Number: MFCD00210626
2
Lithium niobium isopropoxide, 99.9% (metals basis), Thermo Scientific™
CAS: 21864-21-1 Molecular Formula: LiNb(OC3H7)6 MDL Number: MFCD00210626