accessibility menu, dialog, popup

UserName

CAS RN 21816-08-0

IUPAC Name: (5R,7S)-3-bromoadamantane-1-carboxylate
Synonyms: (5R,7S)-3-bromoadamantane-1-carboxylate
Molecular Weight (g/mol): 258.14
Molecular Formula: C11H14BrO2
InChi Key: DJUDQBVINJIMFO-JZVMUCMXSA-M
SMILES: [O-]C(=O)C12C[C@H]3C[C@H](CC(Br)(C3)C1)C2
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)