accessibility menu, dialog, popup

UserName

CAS RN 2181-42-2

IUPAC Name: trimethylsulfanium iodide
Synonyms: sulfonium, trimethyl-, iodide
Molecular Weight (g/mol): 204.07
Molecular Formula: C3H9IS
InChi Key: VFJYIHQDILEQNR-UHFFFAOYSA-M
SMILES: [I-].C[S+](C)C
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)