accessibility menu, dialog, popup

UserName

SeN

CAS RN 2179-79-5

IUPAC Name: (phenylselanyl)formonitrile
Synonyms: (phenylselanyl)formonitrile
Molecular Weight (g/mol): 182.1
Molecular Formula: C7H5NSe
InChi Key: NODWRXQVQYOJGN-UHFFFAOYSA-N
SMILES: N#C[Se]C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results