accessibility menu, dialog, popup

UserName

H3COOHN

CAS RN 21767-12-4

IUPAC Name: 3-(2-methoxyphenoxy)pyrrolidine
Synonyms: 3-(2-methoxyphenoxy)pyrrolidine
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: RLANOFAQNVDAQD-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1OC1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results