accessibility menu, dialog, popup

UserName

CAS RN 21744-83-2

H3CONHO

CAS RN 21744-83-2

IUPAC Name: 2-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 2-methyl-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: PILVMCZSXIINLV-UHFFFAOYNA-N
SMILES: CC1OC2=CC=CC=C2NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results