accessibility menu, dialog, popup

UserName

CAS RN 2170-03-8

CH2OOO

CAS RN 2170-03-8

IUPAC Name: 3-methylideneoxolane-2,5-dione
Synonyms: 3-methylideneoxolane-2,5-dione
Molecular Weight (g/mol): 112.08
Molecular Formula: C5H4O3
InChi Key: OFNISBHGPNMTMS-UHFFFAOYSA-N
SMILES: C=C1CC(=O)OC1=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)