accessibility menu, dialog, popup

UserName

CH3FOH

CAS RN 216976-31-7

IUPAC Name: 3-fluoro-5-methylphenol
Synonyms: 3-fluoro-5-methylphenol
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: AYMBVFCWNCCREA-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results