accessibility menu, dialog, popup

UserName

CAS RN 216144-45-5

IUPAC Name: 1-[4-(4-methylpiperazin-1-yl)phenyl]methanamine
Synonyms: 1-[4-(4-methylpiperazin-1-yl)phenyl]methanamine
Molecular Weight (g/mol): 205.31
Molecular Formula: C12H19N3
InChi Key: MZFQGKRIWIKPBT-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results