CAS RN 2154-32-7
CAS RN 2154-32-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methoxycarbonyl-2,2,5,5-tetramethyl-3-pyrrolidin-1-oxyl, TRC
CAS: 2154-32-7 Chemical Name or Material: 3-Methoxycarbonyl-2,2,5,5-tetramethyl-3-pyrrolidin-1-oxyl Formula Weight: 200.1287 InChI Formula: InChI=1S/C10H18NO3/c1-9(2)6-7(8(12)14-5)10(3,4)11(9)13/h7H,6H2,1-5H3 IUPAC Name: methyl 1-λ^{1}-oxidanyl-2,2,5,5-tetramethylpyrrolidine-3-carboxylate Molecular Formula: C10 H18 N O3 Molecular Weight (g/mol): 200.25 Recommended Storage: -20°C SMILES: COC(=O)C1CC(C)(C)N([O])C1(C)C