accessibility menu, dialog, popup

UserName

CAS RN 21524-34-5

H3CCH3CH3CH3BrH3CCH3

CAS RN 21524-34-5

IUPAC Name: 2-bromo-1,3,5-tris(propan-2-yl)benzene
Synonyms: 2-bromo-1,3,5-triisopropylbenzene
Molecular Weight (g/mol): 283.25
Molecular Formula: C15H23Br
InChi Key: FUMMYHVKFAHQST-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(Br)C(=C1)C(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results