accessibility menu, dialog, popup

UserName

CAS RN 21462-39-5

ClH3C(R)(S)NH3COHNCH3Cl(R)O(R)SCH3(R)OH(S)HO(R)HOH

CAS RN 21462-39-5

IUPAC Name: hydrogen (2S,4R)-N-{2-chloro-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl}-1-methyl-4-propylpyrrolidine-2-carboxamide chloride
Synonyms: hydrogen clindamycin chloride
Molecular Weight (g/mol): 461.44
Molecular Formula: C18H34Cl2N2O5S
InChi Key: AUODDLQVRAJAJM-GQYOKNPUSA-N
SMILES: [H+].[Cl-].CCC[C@@H]1C[C@H](N(C)C1)C(=O)NC(C(C)Cl)[C@H]1O[C@H](SC)[C@H](O)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (4)
  • (9)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (3)
Color 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (9)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (6)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)