accessibility menu, dialog, popup

UserName

NH2H2N

CAS RN 21436-03-3

IUPAC Name: cyclohexane-1,2-diamine
Synonyms: 1,2-cyclohexanediamine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: SSJXIUAHEKJCMH-UHFFFAOYNA-N
SMILES: NC1CCCCC1N
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)