accessibility menu, dialog, popup

UserName

H3COBr

CAS RN 2142-69-0

IUPAC Name: 1-(2-bromophenyl)ethan-1-one
Synonyms: ethanone, 1-(2-bromophenyl)-
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: PIMNFNXBTGPCIL-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1Br
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)