accessibility menu, dialog, popup

UserName

HNOOHOClCl(E/Z)

CAS RN 21395-61-9

IUPAC Name: 3-[(3,4-dichlorophenyl)carbamoyl]prop-2-enoic acid
Synonyms: 3-[(3,4-dichlorophenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 260.07
Molecular Formula: C10H7Cl2NO3
InChi Key: YMYKNYVBUCMJOG-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(=O)NC1=CC(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results