accessibility menu, dialog, popup

UserName

CAS RN 2122196-16-9

SNNNH2NNOOHOHOHSOOHONH3

CAS RN 2122196-16-9

IUPAC Name: 4-[2-({6-amino-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-2-yl}sulfanyl)ethyl]benzene-1-sulfonic acid amine
Synonyms: 4-[2-({6-amino-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-2-yl}sulfanyl)ethyl]benzenesulfonic acid amine
Molecular Weight (g/mol): 500.55
Molecular Formula: C18H24N6O7S2
InChi Key: AYQGFWKUGKSGCZ-UHFFFAOYNA-N
SMILES: N.NC1=C2N=CN(C3OC(CO)C(O)C3O)C2=NC(SCCC2=CC=C(C=C2)S(O)(=O)=O)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results