accessibility menu, dialog, popup

UserName

CAS RN 21221-93-2

FFFFFFO

CAS RN 21221-93-2

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl](phenyl)methanone
Synonyms: [3,5-bis(trifluoromethyl)phenyl](phenyl)methanone
Molecular Weight (g/mol): 318.22
Molecular Formula: C15H8F6O
InChi Key: QETSMUIBVUASOJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)C(=O)C1=CC=CC=C1)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results