accessibility menu, dialog, popup

UserName

CAS RN 211308-81-5

NIH2NCl

CAS RN 211308-81-5

IUPAC Name: 5-chloro-3-iodopyridin-2-amine
Synonyms: 5-chloro-3-iodopyridin-2-amine
Molecular Weight (g/mol): 254.46
Molecular Formula: C5H4ClIN2
InChi Key: DIONPYCYVWCDIG-UHFFFAOYSA-N
SMILES: NC1=C(I)C=C(Cl)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results