accessibility menu, dialog, popup

UserName

CAS RN 2113-57-7

IUPAC Name: 3-bromo-1,1'-biphenyl
Synonyms: 3-bromobiphenyl
Molecular Weight (g/mol): 233.11
Molecular Formula: C12H9Br
InChi Key: USYQKCQEVBFJRP-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results