accessibility menu, dialog, popup

UserName

N

CAS RN 2110-18-1

IUPAC Name: 2-(3-phenylpropyl)pyridine
Synonyms: 2-phenylpropylpyridine
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: JJJPNTQYUJPWGQ-UHFFFAOYSA-N
SMILES: C(CC1=CC=CC=C1)CC1=CC=CC=N1
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)