accessibility menu, dialog, popup

UserName

CAS RN 210963-94-3

CH3OONN

CAS RN 210963-94-3

IUPAC Name: methyl 6-(pyrrolidin-1-yl)pyridine-3-carboxylate
Synonyms: methyl 6-(pyrrolidin-1-yl)pyridine-3-carboxylate
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: OWOKTZZFJFFUKY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(C=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results