accessibility menu, dialog, popup

UserName

CH3OOOCH3

CAS RN 2107-69-9

IUPAC Name: 5,6-dimethoxy-2,3-dihydro-1H-inden-1-one
Synonyms: 5,6-dimethoxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: IHMQOBPGHZFGLC-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=C1OC)C(=O)CC2
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results