accessibility menu, dialog, popup

UserName

OPOOOOPOOOOPOOOOHOPOOOOHNaNaNaNaNaNaNaNa

CAS RN 210488-61-2

IUPAC Name: octasodium 2,6-dihydroxy-3,4,5-tris(phosphonatooxy)cyclohexyl phosphate
Synonyms: octasodium 2,6-dihydroxy-3,4,5-tris(phosphonatooxy)cyclohexyl phosphate
Molecular Weight (g/mol): 675.93
Molecular Formula: C6H8Na8O18P4
InChi Key: QYQMPPDYTHNACQ-UHFFFAOYNA-F
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].OC1C(OP([O-])([O-])=O)C(O)C(OP([O-])([O-])=O)C(OP([O-])([O-])=O)C1OP([O-])([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)